Assay Details
Target Gene Details
Entrez Gene ID: | 10436 |
Gene Name: | EMG1, N1-specific pseudouridine methyltransferase |
Gene Aliases: |
C2F, Grcc2f, NEP1 |
Location: |
Chr.12:6970781-6997428 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EMG1 | NM_001320049.1 | NP_001306978.1 | ||
NM_006331.7 | NP_006322.4 | |||
NR_135131.1 | ||||
AA314580.1 | ||||
AK291597.1 | ||||
AK298207.1 | ||||
AK298745.1 | ||||
AK307177.1 | ||||
BC055314.1 | AAH55314.1 | |||
BM709304.1 | ||||
BP315329.1 | ||||
CD704778.1 | ||||
DA070200.1 | ||||
DC403138.1 | ||||
U56427.1 | ||||
U72514.1 | AAC51641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
