Assay Details
Target Gene Details
Entrez Gene ID: | 5800 |
Gene Name: | protein tyrosine phosphatase, receptor type O |
Gene Aliases: |
GLEPP1, NPHS6, PTP-OC, PTP-U2, PTPROT, PTPU2, R-PTP-O |
Location: |
Chr.12:15322257-15598331 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPRO | NM_002848.3 | NP_002839.1 | ||
NM_030667.2 | NP_109592.1 | |||
XM_017019725.1 | XP_016875214.1 | |||
AB209729.1 | BAD92966.1 | |||
AK289784.1 | ||||
BC035960.1 | AAH35960.1 | |||
BC126201.1 | ||||
BC126203.1 | ||||
DA165109.1 | ||||
DR000351.1 | ||||
U20489.1 | AAA82892.1 | |||
Z48541.1 | CAA88425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33461 | Chr.12:15279453 - 15354673 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
