Assay Details
Target Gene Details
Entrez Gene ID: | 54602 |
Gene Name: | Nedd4 family interacting protein 2 |
Gene Aliases: |
N4WBP5A |
Location: |
Chr.13:79480722-79556077 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDFIP2 | NM_001161407.1 | NP_001154879.1 | ||
| NM_019080.2 | NP_061953.2 | |||
| XM_006719840.3 | XP_006719903.1 | |||
| AB032991.1 | BAA86479.1 | |||
| AB097019.1 | BAC77372.1 | |||
| AB097028.1 | BAC77381.1 | |||
| AB097029.1 | BAC77382.1 | |||
| AB097030.1 | BAC77383.1 | |||
| AB097031.1 | BAC77384.1 | |||
| AK001723.1 | BAA91863.1 | |||
| AK098708.1 | ||||
| AK294839.1 | ||||
| AK297197.1 | ||||
| BC021988.1 | AAH21988.1 | |||
| BC026126.2 | AAH26126.1 | |||
| DC320871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523979 | Chr.13:79405544 - 79546128 on Build GRCh38 | Loss |
|
| esv3632819 | Chr.13:79393219 - 79532503 on Build GRCh38 | Gain |
|
| nsv832657 | Chr.13:79330843 - 79524973 on Build GRCh38 | Gain |
|
| esv3632814 | Chr.13:79199135 - 79520919 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117515623] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map