Assay Details
Target Gene Details
Entrez Gene ID: | 2971 |
Gene Name: | general transcription factor IIIA |
Gene Aliases: |
AP2, TFIIIA |
Location: |
Chr.13:27424544-27435709 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3A | NM_002097.2 | NP_002088.2 | ||
| AK057993.1 | ||||
| BP254755.1 | ||||
| D32257.1 | BAA06988.1 | |||
| U14134.1 | AAA21873.1 | |||
| U20272.1 | AAA75623.1 |
More Information
Set Membership: |
|

Genomic Map