Assay Details
Target Gene Details
Entrez Gene ID: | 145581 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 5 |
Gene Aliases: |
C14orf146, FIGLER8, SALM5 |
Location: |
Chr.14:41604684-41904549 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRFN5 | NM_152447.3 | NP_689660.2 | ||
XM_005267369.3 | XP_005267426.1 | |||
XM_017021049.1 | XP_016876538.1 | |||
XM_017021050.1 | XP_016876539.1 | |||
AK055365.1 | BAB70910.1 | |||
AK096627.1 | ||||
AK299632.1 | ||||
BC043165.1 | AAH43165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759983 | Chr.14:41576670 - 41790308 on Build GRCh38 | Loss |
![]() |
nsv1042078 | Chr.14:41524782 - 41650940 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
