Assay Details
Target Gene Details
Entrez Gene ID: | 197135 |
Gene Name: | PAT1 homolog 2 |
Gene Aliases: |
Pat1a, hPat1a |
Location: |
Chr.15:44665732-44711389 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PATL2 | NM_001145112.1 | NP_001138584.1 | ||
| XM_011521336.2 | XP_011519638.2 | |||
| XM_011521337.2 | XP_011519639.2 | |||
| XM_011521338.2 | XP_011519640.1 | |||
| XM_011521339.2 | XP_011519641.1 | |||
| XM_011521340.2 | XP_011519642.1 | |||
| XM_011521341.1 | XP_011519643.1 | |||
| XM_011521342.2 | XP_011519644.1 | |||
| XM_011521343.2 | XP_011519645.1 | |||
| XM_011521344.2 | XP_011519646.1 | |||
| XM_011521345.2 | XP_011519647.1 | |||
| XM_011521346.2 | XP_011519648.2 | |||
| XM_011521347.1 | XP_011519649.1 | |||
| XM_011521348.2 | XP_011519650.1 | |||
| XM_017022000.1 | XP_016877489.1 | |||
| XM_017022001.1 | XP_016877490.1 | |||
| BI909158.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1517 | Chr.15:44662336 - 44697647 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map