Assay Details
Target Gene Details
Entrez Gene ID: | 1854 |
Gene Name: | deoxyuridine triphosphatase |
Gene Aliases: |
dUTPase |
Location: |
Chr.15:48331018-48343373 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUT | NM_001025248.1 | NP_001020419.1 | ||
| NM_001025249.1 | NP_001020420.1 | |||
| NM_001948.3 | NP_001939.1 | |||
| XM_011521315.2 | XP_011519617.1 | |||
| XM_017021988.1 | XP_016877477.1 | |||
| AB049113.2 | BAB13724.1 | |||
| AK000629.1 | ||||
| AK291515.1 | ||||
| AK298464.1 | ||||
| AK312122.1 | ||||
| BC033645.1 | AAH33645.1 | |||
| BC070339.1 | AAH70339.1 | |||
| BC110377.1 | ||||
| BG505144.1 | ||||
| CR541720.1 | CAG46521.1 | |||
| CR541781.1 | CAG46580.1 | |||
| L11877.1 | AAA36801.1 | |||
| M89913.1 | AAA58444.1 | |||
| U31930.1 | AAC50418.1 | |||
| U62891.1 | AAC51123.1 | |||
| U90223.1 | AAB94642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636412 | Chr.15:48322023 - 48347079 on Build GRCh38 | Gain |
|
| esv2760030 | Chr.15:48216841 - 48371581 on Build GRCh38 | Loss |
|
| esv3569565 | Chr.15:48317122 - 48346517 on Build GRCh38 | Gain |
|
| nsv1053885 | Chr.15:48314149 - 48346517 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map