Assay Details
Target Gene Details
Entrez Gene ID: | 22801 |
Gene Name: | integrin subunit alpha 11 |
Gene Aliases: |
HsT18964 |
Location: |
Chr.15:68297433-68432312 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA11 | XM_005254228.3 | 28 | 8157 | XP_005254285.1 |
| XM_011521363.2 | 28 | 7887 | XP_011519665.1 | |
| AL117454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749815 | Chr.15:68298316 - 68299660 on Build GRCh38 | Deletion |
|
| esv2760036 | Chr.15:68245734 - 68522324 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map