Assay Details
Target Gene Details
Entrez Gene ID: | 654364 |
Gene Name: | NME1-NME2 readthrough |
Gene Aliases: |
NM23-LV, NMELV |
Location: |
Chr.17:51153536-51171744 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NME1-NME2 | NM_001018136.2 | NP_001018146.1 | ||
NR_037149.1 | ||||
AK313448.1 | ||||
BC107894.1 | AAI07895.1 | |||
BC133029.1 | ||||
BC133031.1 | ||||
BI603164.1 | ||||
DQ109675.1 | AAZ82097.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
