Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641599 | Chr.18:3504024 - 3507639 on Build GRCh38 | Loss |
|
| nsv1059696 | Chr.18:3494893 - 3572306 on Build GRCh38 | Gain |
|
| dgv52n6 | Chr.18:3502646 - 3508570 on Build GRCh38 | Deletion |
|
| dgv577e199 | Chr.18:3504050 - 3507531 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map