Assay Details
Target Gene Details
Entrez Gene ID: | 66002 |
Gene Name: | cytochrome P450 family 4 subfamily F member 12 |
Gene Aliases: |
CYPIVF12, F22329_1 |
Location: |
Chr.19:15673018-15698819 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4F12 | NM_023944.3 | NP_076433.3 | ||
| NR_117085.1 | ||||
| XM_006722850.3 | XP_006722913.1 | |||
| XM_011528202.2 | XP_011526504.1 | |||
| XM_011528203.2 | XP_011526505.1 | |||
| XM_011528204.2 | XP_011526506.1 | |||
| XM_011528205.2 | XP_011526507.1 | |||
| XM_011528207.2 | XP_011526509.1 | |||
| XM_011528208.2 | XP_011526510.1 | |||
| XM_017027172.1 | XP_016882661.1 | |||
| AB035130.1 | BAB18269.1 | |||
| AB035131.1 | BAB18270.1 | |||
| AK074412.1 | ||||
| AK075435.1 | ||||
| AL832171.1 | CAI46131.1 | |||
| AY008841.1 | AAG33247.1 | |||
| AY358977.1 | AAQ89336.1 | |||
| BC035350.1 | AAH35350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470128 | Chr.19:15653261 - 15728555 on Build GRCh38 | Gain |
|
| esv2421519 | Chr.19:15668643 - 15719658 on Build GRCh38 | Duplication |
|
| esv2758747 | Chr.19:15669231 - 15740451 on Build GRCh38 | Gain+Loss |
|
| esv3334671 | Chr.19:15673766 - 15931354 on Build GRCh38 | Duplication |
|
| nsv978781 | Chr.19:15682273 - 15685174 on Build GRCh38 | Duplication |
|
| dgv28n64 | Chr.19:15669207 - 15719658 on Build GRCh38 | Gain |
|
| dgv3452n100 | Chr.19:15679275 - 15714929 on Build GRCh38 | Gain |
|
| dgv3446n100 | Chr.19:15661423 - 15714929 on Build GRCh38 | Gain |
|
| nsv9671 | Chr.19:15666486 - 15723022 on Build GRCh38 | Gain |
|
| esv2760506 | Chr.19:15666091 - 15726410 on Build GRCh38 | Gain+Loss |
|
| esv3643776 | Chr.19:15672308 - 15725395 on Build GRCh38 | Gain |
|
| nsv436858 | Chr.19:15670801 - 15720751 on Build GRCh38 | Insertion |
|
| esv22534 | Chr.19:15667246 - 15726197 on Build GRCh38 | Gain+Loss |
|
| dgv6289n54 | Chr.19:15663105 - 15724712 on Build GRCh38 | Gain |
|
| nsv526168 | Chr.19:15669937 - 15748902 on Build GRCh38 | Gain |
|
| nsv1064684 | Chr.19:15311319 - 16062011 on Build GRCh38 | Gain |
|
| dgv137e55 | Chr.19:15666599 - 15728649 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115854510] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map