Assay Details
Target Gene Details
Entrez Gene ID: | 27134 |
Gene Name: | tight junction protein 3 |
Gene Aliases: |
ZO-3, ZO3 |
Location: |
Chr.19:3708337-3750813 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TJP3 | NM_001267560.1 | NP_001254489.1 | ||
| AK091118.1 | ||||
| AK304462.1 | ||||
| AL555606.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509718 | Chr.19:3676947 - 3844683 on Build GRCh38 | Insertion |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map