Assay Details
Target Gene Details
Entrez Gene ID: | 3640 |
Gene Name: | insulin like 3 |
Gene Aliases: |
RLF, RLNL, ley-I-L |
Location: |
Chr.19:17816513-17821574 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INSL3 | NM_001265587.1 | NP_001252516.1 | ||
| NM_005543.3 | NP_005534.2 | |||
| AK302780.1 | ||||
| AY082014.1 | AAL92559.1 | |||
| BC032810.1 | AAH32810.1 | |||
| BC053345.1 | AAH53345.1 | |||
| BC071706.1 | AAH71706.1 | |||
| BC106721.1 | AAI06722.1 | |||
| BC106722.1 | AAI06723.1 | |||
| BX334591.2 | ||||
| S72482.1 | AAB31371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv50n68 | Chr.19:17671882 - 17842923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map