Assay Details
Target Gene Details
Entrez Gene ID: | 101927377 |
Gene Name: | uncharacterized LOC101927377 |
Gene Aliases: |
- |
Location: |
Chr.20:47352561-47354633 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927377 | NR_134501.1 |
Target Gene Details
Entrez Gene ID: | 23613 |
Gene Name: | zinc finger MYND-type containing 8 |
Gene Aliases: |
PRKCBP1, PRO2893, RACK7 |
Location: |
Chr.20:47209214-47357780 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33497 | Chr.20:47143439 - 47650962 on Build GRCh38 | Loss |
|
| nsv1066814 | Chr.20:47342953 - 47580758 on Build GRCh38 | Gain |
|
| nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
|
| nsv1066907 | Chr.20:47310827 - 47736827 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map