Assay Details
Target Gene Details
Entrez Gene ID: | 114041 |
Gene Name: | B3GALT5 antisense RNA 1 |
Gene Aliases: |
C21orf88 |
Location: |
Chr.21:39597147-39612822 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GALT5-AS1 | NR_026542.1 | |||
| NR_026543.1 | ||||
| AF426266.1 | AAM53522.1 | |||
| AF426267.1 | AAM53523.1 | |||
| BG751912.1 | ||||
| DA218783.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27418 | Chr.21:39609713 - 39615556 on Build GRCh38 | Gain |
|
| esv3647020 | Chr.21:39531228 - 39615172 on Build GRCh38 | Gain |
|
| esv3568195 | Chr.21:39608433 - 39615629 on Build GRCh38 | Loss |
|
| nsv1061454 | Chr.21:39386047 - 39893479 on Build GRCh38 | Gain |
|

Genomic Map