Assay Details
Target Gene Details
Entrez Gene ID: | 114041 |
Gene Name: | B3GALT5 antisense RNA 1 |
Gene Aliases: |
C21orf88 |
Location: |
Chr.21:39597147-39612822 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GALT5-AS1 | NR_026542.1 | |||
| AF426266.1 | AAM53522.1 | |||
| DA218783.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34779 | Chr.21:39512944 - 39605546 on Build GRCh38 | Gain |
|
| esv3647020 | Chr.21:39531228 - 39615172 on Build GRCh38 | Gain |
|
| nsv1061454 | Chr.21:39386047 - 39893479 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75380972] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map