Assay Details
Target Gene Details
Entrez Gene ID: | 3761 |
Gene Name: | potassium voltage-gated channel subfamily J member 4 |
Gene Aliases: |
HIR, HIRK2, HRK1, IRK-3, IRK3, Kir2.3 |
Location: |
Chr.22:38426327-38455199 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNJ4 | NM_004981.1 | NP_004972.1 | ||
NM_152868.2 | NP_690607.1 | |||
BC032562.1 | AAH32562.1 | |||
CR456507.1 | CAG30393.1 | |||
S72503.1 | AAC60632.1 | |||
U07364.1 | AAA19962.1 | |||
U24056.1 | AAA66076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834195 | Chr.22:38300366 - 38490431 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060077 | Chr.22:38419538 - 38460822 on Build GRCh38 | Gain |
![]() ![]() |
nsv817994 | Chr.22:38413480 - 38469858 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1058338 | Chr.22:38423344 - 38450414 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
