Assay Details
Target Gene Details
Entrez Gene ID: | 2747 |
Gene Name: | glutamate dehydrogenase 2 |
Gene Aliases: |
GDH2, GLUDP1 |
Location: |
Chr.X:121047608-121049942 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLUD2 | NM_012084.3 | 1 | 1625 | NP_036216.2 |
| AK313356.1 | 1 | 1622 | ||
| BC005111.1 | 1 | 668 | AAH05111.1 | |
| BC050732.2 | 1 | 1625 | AAH50732.1 |
More Information
Set Membership: |
|

Genomic Map