Assay Details
Target Gene Details
Entrez Gene ID: | 6421 |
Gene Name: | splicing factor proline and glutamine rich |
Gene Aliases: |
POMP100, PPP1R140, PSF |
Location: |
Chr.1:35176378-35193161 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFPQ | NR_136702.1 | |||
| NR_136703.1 | ||||
| XM_005271112.4 | XP_005271169.1 | |||
| XM_005271113.4 | XP_005271170.1 | |||
| XM_005271115.4 | XP_005271172.1 | |||
| XM_011541950.2 | XP_011540252.1 | |||
| XM_017002053.1 | XP_016857542.1 | |||
| XM_017002054.1 | XP_016857543.1 | |||
| AK095702.1 | 1 | 1777 | ||
| AL831889.1 | ||||
| AL833670.1 | ||||
| BX537811.1 |
More Information
Set Membership: |
|

Genomic Map