Assay Details
Target Gene Details
Entrez Gene ID: | 829 |
Gene Name: | capping actin protein of muscle Z-line alpha subunit 1 |
Gene Aliases: |
CAPPA1, CAPZ, CAZ1 |
Location: |
Chr.1:112619453-112671619 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZA1 | NM_006135.2 | NP_006126.1 | ||
| XM_011542225.2 | XP_011540527.1 | |||
| XM_017002424.1 | XP_016857913.1 | |||
| AK223226.1 | BAD96946.1 | |||
| AK289654.1 | ||||
| BC000144.1 | AAH00144.1 | |||
| BT019364.1 | AAV38171.1 | |||
| BX648738.1 | ||||
| CR407657.1 | CAG28585.1 | |||
| CR541819.1 | CAG46618.1 | |||
| U56637.1 | AAC00533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27061 | Chr.1:112150007 - 112703641 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map