Assay Details
Target Gene Details
Entrez Gene ID: | 6706 |
Gene Name: | small proline rich protein 2G |
Gene Aliases: |
- |
Location: |
Chr.1:153149582-153195377 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRR2G | XM_017002177.1 | XP_016857666.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547899 | Chr.1:152699529 - 153272606 on Build GRCh38 | Gain |
|
| nsv1004168 | Chr.1:153115733 - 153253557 on Build GRCh38 | Loss |
|
| nsv2932 | Chr.1:153172965 - 153206537 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map