Assay Details
Target Gene Details
Entrez Gene ID: | 51154 |
Gene Name: | MRT4 homolog, ribosome maturation factor |
Gene Aliases: |
C1orf33, MRT4, dJ657E11.4 |
Location: |
Chr.1:19251581-19260128 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRTO4 | NM_016183.3 | 8 | 1873 | NP_057267.2 |
| XM_006710675.3 | 8 | 2031 | XP_006710738.1 |
More Information
Set Membership: |
|

Genomic Map