Assay Details
Target Gene Details
Entrez Gene ID: | 84950 |
Gene Name: | pre-mRNA processing factor 38A |
Gene Aliases: |
Prp38 |
Location: |
Chr.1:52404547-52419723 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF38A | XM_011542315.2 | 10 | 4050 | XP_011540617.1 |
More Information
Set Membership: |
|

Genomic Map