Assay Details
Target Gene Details
Entrez Gene ID: | 55924 |
Gene Name: | family with sequence similarity 212 member B |
Gene Aliases: |
C1orf183 |
Location: |
Chr.1:111722064-111755797 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM212B | NM_198926.2 | NP_945120.1 | ||
AK293989.1 | ||||
AL137199.1 | CAB69909.1 |
Target Gene Details
Entrez Gene ID: | 100506343 |
Gene Name: | FAM212B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:111739841-111747798 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM212B-AS1 | NR_038951.1 | |||
NR_038952.1 | ||||
AK092511.1 | ||||
BG329450.1 |
