Assay Details
Target Gene Details
Entrez Gene ID: | 3749 |
Gene Name: | potassium voltage-gated channel subfamily C member 4 |
Gene Aliases: |
C1orf30, HKSHIIIC, KSHIIIC, KV3.4 |
Location: |
Chr.1:110210714-110282649 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNC4 | NM_001039574.2 | NP_001034663.1 | ||
| NM_004978.4 | NP_004969.2 | |||
| NR_036437.1 | ||||
| XM_006710625.3 | XP_006710688.1 | |||
| XM_006710627.3 | XP_006710690.1 | |||
| XM_011541401.2 | XP_011539703.1 | |||
| XM_011541402.1 | XP_011539704.1 | |||
| XM_011541403.1 | XP_011539705.1 | |||
| XM_011541404.1 | XP_011539706.1 | |||
| AK310193.1 | ||||
| BX098184.1 | ||||
| BX537638.1 | 1 | 4846 | ||
| CK823243.1 | ||||
| DA936036.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1075780 | Chr.1:110221577 - 110226178 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map