Assay Details
Target Gene Details
Entrez Gene ID: | 9462 |
Gene Name: | RAS protein activator like 2 |
Gene Aliases: |
NGAP |
Location: |
Chr.1:178093729-178479514 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASAL2 | NM_170692.2 | NP_733793.2 | ||
| XM_011510166.2 | XP_011508468.1 | |||
| XM_011510167.2 | XP_011508469.1 | |||
| AB209789.1 | BAD93026.1 | |||
| AK075169.1 | ||||
| AK311017.1 | ||||
| AV706305.1 | ||||
| BC063442.1 | AAH63442.1 | |||
| BC078167.1 | AAH78167.1 | |||
| BC110611.2 | AAI10612.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548252 | Chr.1:177758922 - 178428784 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map