Assay Details
Target Gene Details
Entrez Gene ID: | 10806 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP |
Location: |
Chr.1:243255328-243500092 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001488 | Chr.1:243227523 - 243419972 on Build GRCh38 | Loss |
|
| nsv517939 | Chr.1:243213455 - 243359604 on Build GRCh38 | Gain |
|
| esv32774 | Chr.1:242878641 - 243304780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map