Assay Details
Target Gene Details
Entrez Gene ID: | 128338 |
Gene Name: | DNA damage regulated autophagy modulator 2 |
Gene Aliases: |
CORD21, PRO180, TMEM77, WWFQ154 |
Location: |
Chr.1:111117332-111140216 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRAM2 | NM_178454.4 | NP_848549.3 | ||
| XM_005270469.2 | XP_005270526.1 | |||
| XM_005270470.1 | XP_005270527.1 | |||
| XM_006710361.1 | XP_006710424.1 | |||
| XM_006710362.1 | XP_006710425.1 | |||
| XM_011540707.1 | XP_011539009.1 | |||
| XM_011540708.1 | XP_011539010.1 | |||
| XM_017000320.1 | XP_016855809.1 | |||
| XM_017000321.1 | XP_016855810.1 | |||
| AK075350.1 | BAC11562.1 | |||
| AK309557.1 | ||||
| AL833098.1 | ||||
| AY336747.1 | AAR02410.1 | |||
| AY358492.1 | AAQ88856.1 | |||
| BC047025.1 | AAH47025.3 | |||
| BC091509.1 | AAH91509.1 | |||
| EF710624.1 |
More Information
Set Membership: |
|

Genomic Map