Assay Details
Target Gene Details
Entrez Gene ID: | 9295 |
Gene Name: | serine and arginine rich splicing factor 11 |
Gene Aliases: |
NET2, SFRS11, dJ677H15.2, p54 |
Location: |
Chr.1:70205682-70252020 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF11 | NM_001190987.1 | NP_001177916.1 | ||
| NM_004768.3 | NP_004759.1 | |||
| XM_005271338.3 | XP_005271395.1 | |||
| XM_005271339.3 | XP_005271396.1 | |||
| XM_011542431.2 | XP_011540733.1 | |||
| XM_017002811.1 | XP_016858300.1 | |||
| XM_017002812.1 | XP_016858301.1 | |||
| XM_017002813.1 | XP_016858302.1 | |||
| XM_017002816.1 | XP_016858305.1 | |||
| XM_017002824.1 | XP_016858313.1 | |||
| XM_017002828.1 | XP_016858317.1 | |||
| XM_017002829.1 | XP_016858318.1 | |||
| XM_017002830.1 | XP_016858319.1 | |||
| AK300147.1 | ||||
| AK310447.1 | ||||
| AK310666.1 | ||||
| AK311184.1 | ||||
| BC032727.1 | ||||
| BC040436.1 | AAH40436.1 | |||
| M74002.1 | AAA35554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1343 | Chr.1:70199486 - 70231317 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map