Assay Details
Target Gene Details
| Entrez Gene ID: | 80135 | 
| Gene Name: | ribosome production factor 1 homolog | 
| Gene Aliases: | BXDC5 | 
| Location: | Chr.1:84479237-84498350 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| RPF1 | NM_025065.6 | NP_079341.2 | ||
| AF322053.1 | AAL37236.1 | |||
| AK022537.1 | BAB14086.1 | |||
| AL359584.2 | CAH10671.1 | |||
| BC016051.1 | AAH16051.1 | |||
| CR627468.1 | CAH10670.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv830437 | Chr.1:84371841 - 84535586 on Build GRCh38 | Gain+Loss |  UOX  DNASE2B  SPATA1  RPF1  GNG5 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top