Assay Details
Target Gene Details
Entrez Gene ID: | 55160 |
Gene Name: | Rho guanine nucleotide exchange factor 10 like |
Gene Aliases: |
GrinchGEF |
Location: |
Chr.1:17520197-17697875 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF10L | XM_006710728.1 | XP_006710791.1 | ||
| XM_006710731.1 | XP_006710794.1 | |||
| AK129959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4187 | Chr.1:17503448 - 17537498 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114495464] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map