Assay Details
Target Gene Details
Entrez Gene ID: | 127534 |
Gene Name: | gap junction protein beta 4 |
Gene Aliases: |
CX30.3, EKV |
Location: |
Chr.1:34759741-34763724 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB4 | NM_153212.2 | NP_694944.1 | ||
| XM_011540679.2 | XP_011538981.1 | |||
| AK057628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv947426 | Chr.1:34760525 - 34782365 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map