Assay Details
Target Gene Details
Entrez Gene ID: | 7388 |
Gene Name: | ubiquinol-cytochrome c reductase hinge protein |
Gene Aliases: |
QCR6, UQCR8 |
Location: |
Chr.1:46303613-46316777 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UQCRH | NM_001297565.1 | NP_001284494.1 | ||
| NM_001297566.1 | NP_001284495.1 | |||
| NM_006004.3 | NP_005995.2 | |||
| AA526171.1 | ||||
| AK311967.1 | ||||
| BC001426.2 | AAH01426.1 | |||
| BC001934.1 | AAH01934.1 | |||
| BC015177.1 | AAH15177.1 | |||
| BC093060.1 | AAH93060.1 | |||
| BC107703.1 | AAI07704.1 | |||
| BG472687.1 | ||||
| BP366403.1 | ||||
| BT007070.1 | AAP35733.1 | |||
| M36647.1 | AAA36317.1 | |||
| Y00764.1 | CAA68733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829748 | Chr.1:46258836 - 46410450 on Build GRCh38 | Loss |
|
| nsv470711 | Chr.1:46194623 - 46344858 on Build GRCh38 | Gain |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map