Assay Details
Target Gene Details
Entrez Gene ID: | 387338 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 4 |
Gene Aliases: |
SHTAP |
Location: |
Chr.1:46340177-46365152 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN4 | NM_001256127.1 | NP_001243056.1 | ||
| NM_001256128.1 | NP_001243057.1 | |||
| NM_199044.3 | NP_950245.2 | |||
| NR_045789.1 | ||||
| NR_045790.1 | ||||
| NR_045791.1 | ||||
| AK021577.1 | BAB13847.1 | |||
| AK057420.1 | ||||
| AK097524.1 | ||||
| AK128066.1 | BAC87258.1 | |||
| AK291741.1 | ||||
| AK292353.1 | ||||
| AK295003.1 | ||||
| BC014441.1 | AAH14441.1 | |||
| BX360729.2 | ||||
| BX648592.1 | ||||
| DA799314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577837 | Chr.1:46338361 - 46357126 on Build GRCh38 | Loss |
|
| nsv829748 | Chr.1:46258836 - 46410450 on Build GRCh38 | Loss |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
| nsv546162 | Chr.1:46341925 - 46405089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map