Assay Details
Target Gene Details
Entrez Gene ID: | 481 |
Gene Name: | ATPase Na+/K+ transporting subunit beta 1 |
Gene Aliases: |
ATP1B |
Location: |
Chr.1:169106709-169132722 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP1B1 | NM_001677.3 | NP_001668.1 | ||
| AK290181.1 | ||||
| AK296059.1 | ||||
| AK316043.1 | ||||
| AY545986.1 | ||||
| BC000006.2 | AAH00006.1 | |||
| BQ934151.1 | ||||
| BT009787.1 | AAP88789.1 | |||
| CR407616.1 | CAG28544.1 | |||
| U16799.1 | AAC50132.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss |
|
| nsv831858 | Chr.1:169053145 - 169209830 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map