Assay Details
Target Gene Details
Entrez Gene ID: | 9372 |
Gene Name: | zinc finger FYVE-type containing 9 |
Gene Aliases: |
MADHIP, NSP, PPP1R173, SARA, SMADIP |
Location: |
Chr.1:52142001-52348664 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFYVE9 | NM_004799.3 | NP_004790.2 | ||
| NM_007324.3 | NP_015563.2 | |||
| XM_017002845.1 | XP_016858334.1 | |||
| AF130419.1 | AAD31694.1 | |||
| AF130420.1 | AAD31695.1 | |||
| DR005493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv855 | Chr.1:52155519 - 52187874 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72893911] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map