Assay Details
Target Gene Details
Entrez Gene ID: | 6262 |
Gene Name: | ryanodine receptor 2 |
Gene Aliases: |
ARVC2, ARVD2, RYR-2, RyR, VTSIP |
Location: |
Chr.1:237042208-237833988 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RYR2 | NM_001035.2 | NP_001026.2 | ||
| XM_006711802.3 | XP_006711865.1 | |||
| XM_006711803.3 | XP_006711866.1 | |||
| XM_006711804.3 | XP_006711867.1 | |||
| XM_006711805.3 | XP_006711868.1 | |||
| XM_006711806.3 | XP_006711869.1 | |||
| XM_006711807.3 | XP_006711870.1 | |||
| XM_006711808.3 | XP_006711871.1 | |||
| XM_006711809.3 | XP_006711872.1 | |||
| XM_006711810.3 | XP_006711873.1 | |||
| XM_017002028.1 | XP_016857517.1 | |||
| X98330.1 | CAA66975.1 | |||
| Y08218.1 | CAA69395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013771 | Chr.1:237064443 - 237163179 on Build GRCh38 | Loss |
|
| nsv1000707 | Chr.1:237019112 - 237359712 on Build GRCh38 | Gain |
|
| nsv954001 | Chr.1:237070201 - 237076200 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map