Assay Details
Target Gene Details
Entrez Gene ID: | 83593 |
Gene Name: | Ras association domain family member 5 |
Gene Aliases: |
Maxp1, NORE1, NORE1A, NORE1B, RAPL, RASSF3 |
Location: |
Chr.1:206507530-206589284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF5 | NM_182663.3 | NP_872604.1 | ||
| NM_182664.3 | NP_872605.1 | |||
| AB209145.1 | BAD92382.1 | |||
| AK289861.1 | ||||
| AK291268.1 | ||||
| AY062002.1 | AAL40388.1 | |||
| AY062003.1 | AAL40389.1 | |||
| BC004270.1 | AAH04270.1 | |||
| BC007203.1 | AAH07203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2672106 | Chr.1:206522812 - 206525659 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map