Assay Details
Target Gene Details
Entrez Gene ID: | 9859 |
Gene Name: | centrosomal protein 170 |
Gene Aliases: |
FAM68A, KAB, KIAA0470 |
Location: |
Chr.1:243124428-243255936 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517939 | Chr.1:243213455 - 243359604 on Build GRCh38 | Gain |
|
| esv32774 | Chr.1:242878641 - 243304780 on Build GRCh38 | Gain |
|
| nsv1001378 | Chr.1:243144352 - 243222557 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map