Assay Details
Target Gene Details
Entrez Gene ID: | 5500 |
Gene Name: | protein phosphatase 1 catalytic subunit beta |
Gene Aliases: |
HEL-S-80p, PP-1B, PP1B, PP1beta, PPP1CD |
Location: |
Chr.2:28751748-28802940 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1CB | NM_002709.2 | NP_002700.1 | ||
| NM_206876.1 | NP_996759.1 | |||
| AF092905.1 | AAF01137.1 | |||
| AK295957.1 | ||||
| AK303682.1 | ||||
| AK312329.1 | ||||
| AK316532.1 | ||||
| BC002697.2 | AAH02697.1 | |||
| BC012045.1 | AAH12045.1 | |||
| BI551898.1 | ||||
| BT019744.1 | AAV38549.1 | |||
| BX477003.1 | ||||
| BX647295.1 | ||||
| BX647970.1 | ||||
| CD656807.1 | ||||
| CR542263.1 | CAG47059.1 | |||
| CR542285.1 | CAG47080.1 | |||
| EU668339.1 | ||||
| X80910.1 | CAA56870.1 |
More Information
Set Membership: |
|

Genomic Map