Assay Details
Target Gene Details
| Entrez Gene ID: | 105373805 | 
| Gene Name: | uncharacterized LOC105373805 | 
| Gene Aliases: | - | 
| Location: | Chr.2:191021526-191032314 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LOC105373805 | NR_136318.1 | |||
| AW873644.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv999391 | Chr.2:190884309 - 191057205 on Build GRCh38 | Gain |  STAT4  LOC105373805  GLS  STAT1 | 
| nsv834493 | Chr.2:190946647 - 191109514 on Build GRCh38 | Gain |  STAT4  LOC105373805  GLS  STAT1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic