Assay Details
Target Gene Details
Entrez Gene ID: | 2571 |
Gene Name: | glutamate decarboxylase 1 |
Gene Aliases: |
CPSQ1, GAD, SCP |
Location: |
Chr.2:170813210-170861151 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAD1 | NM_000817.2 | NP_000808.2 | ||
| NM_013445.3 | NP_038473.2 | |||
| XM_005246444.2 | XP_005246501.1 | |||
| XM_011510922.1 | XP_011509224.1 | |||
| XM_017003756.1 | XP_016859245.1 | |||
| XM_017003757.1 | XP_016859246.1 | |||
| XM_017003758.1 | XP_016859247.1 | |||
| A28074.1 | CAA01913.1 | |||
| AF178853.2 | AAF18390.2 | |||
| AK292582.1 | ||||
| BC002815.2 | AAH02815.1 | |||
| BC026349.1 | AAH26349.1 | |||
| BC036552.1 | AAH36552.1 | |||
| BC037780.2 | AAH37780.1 | |||
| KU178086.1 | ||||
| KU178087.1 | ||||
| KU178088.1 | ||||
| L16888.1 | AAB59427.1 | |||
| M81883.1 | AAA62368.1 | |||
| S61897.1 | AAB26937.1 | |||
| S61898.1 | AAB26938.1 | |||
| Z22750.1 | CAA80435.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528929 | Chr.2:170815681 - 170821869 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map