Assay Details
Target Gene Details
Entrez Gene ID: | 10097 |
Gene Name: | ARP2 actin related protein 2 homolog |
Gene Aliases: |
ARP2 |
Location: |
Chr.2:65227695-65271253 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR2 | NM_001005386.2 | NP_001005386.1 | ||
| NM_005722.3 | NP_005713.1 | |||
| AF006082.1 | AAB64187.1 | |||
| AK295157.1 | ||||
| AK301634.1 | ||||
| AK315205.1 | ||||
| BC014546.1 | AAH14546.1 | |||
| BC036253.1 | AAH36253.1 | |||
| BI670123.1 | ||||
| BX491447.1 | ||||
| BX649080.1 | ||||
| DC404298.1 |
More Information
Set Membership: |
|

Genomic Map