Assay Details
Target Gene Details
Entrez Gene ID: | 2820 |
Gene Name: | glycerol-3-phosphate dehydrogenase 2 |
Gene Aliases: |
GDH2, GPDM, mGPDH |
Location: |
Chr.2:156435290-156586403 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPD2 | NM_000408.4 | NP_000399.3 | ||
| NM_001083112.2 | NP_001076581.2 | |||
| XM_011510977.2 | XP_011509279.1 | |||
| XM_017003830.1 | XP_016859319.1 | |||
| XM_017003831.1 | XP_016859320.1 | |||
| AB209399.1 | BAD92636.1 | |||
| AK290057.1 | ||||
| AK291065.1 | ||||
| AK292817.1 | ||||
| AK299645.1 | ||||
| AL699359.1 | ||||
| BC019874.1 | AAH19874.1 | |||
| U12424.1 | AAA65701.1 | |||
| U36310.1 | AAB60403.1 | |||
| U79250.1 | AAB50200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3593037 | Chr.2:156441659 - 156443069 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map