Assay Details
Target Gene Details
Entrez Gene ID: | 10408 |
Gene Name: | MYCN opposite strand |
Gene Aliases: |
MYCN-AS1, N-CYM, NCYM, NYCM |
Location: |
Chr.2:15936265-15941723 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCNOS | NR_110230.1 | 4 | 794 |
More Information
Set Membership: |
|

Genomic Map