Assay Details
Target Gene Details
Entrez Gene ID: | 101927027 |
Gene Name: | uncharacterized LOC101927027 |
Gene Aliases: |
- |
Location: |
Chr.2:178413659-178440243 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927027 | NR_110204.1 | |||
| NR_110205.1 | ||||
| NR_110206.1 | ||||
| AL520497.3 | ||||
| BI915242.1 | ||||
| BX538254.1 | ||||
| DA520566.1 | ||||
| DT216517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834466 | Chr.2:178276065 - 178420051 on Build GRCh38 | Gain |
|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|

Genomic Map