Assay Details
Target Gene Details
Entrez Gene ID: | 90134 |
Gene Name: | potassium voltage-gated channel subfamily H member 7 |
Gene Aliases: |
ERG3, HERG3, Kv11.3 |
Location: |
Chr.2:162371407-162838747 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH7 | NM_033272.3 | NP_150375.2 | ||
| NM_173162.2 | NP_775185.1 | |||
| XM_011512109.2 | XP_011510411.1 | |||
| XM_017005218.1 | XP_016860707.1 | |||
| XM_017005219.1 | XP_016860708.1 | |||
| XM_017005220.1 | XP_016860709.1 | |||
| XM_017005221.1 | XP_016860710.1 | |||
| AF032897.1 | AAD01946.1 | |||
| BC035815.1 | AAH35815.1 | |||
| DR002425.1 |
Target Gene Details
Entrez Gene ID: | 101929570 |
Gene Name: | uncharacterized LOC101929570 |
Gene Aliases: |
- |
Location: |
Chr.2:162768936-162797972 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929570 | NR_110258.2 |
More Information
Set Membership: |
|

Genomic Map