Assay Details
Target Gene Details
Entrez Gene ID: | 390 |
Gene Name: | Rho family GTPase 3 |
Gene Aliases: |
ARHE, Rho8, RhoE, memB |
Location: |
Chr.2:150468193-150487695 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RND3 | NM_001254738.1 | NP_001241667.1 | ||
| NM_005168.4 | NP_005159.1 | |||
| AF498969.1 | AAM21116.1 | |||
| AK298007.1 | ||||
| AK299731.1 | ||||
| AK309442.1 | ||||
| AK311312.1 | ||||
| AK313224.1 | ||||
| AL702945.1 | ||||
| BC012513.1 | AAH12513.1 | |||
| BQ440721.1 | ||||
| BT006769.1 | AAP35415.1 | |||
| CR542038.1 | CAG46835.1 | |||
| CR542070.1 | CAG46867.1 | |||
| DC367501.1 | ||||
| S82240.1 | AAB47133.1 | |||
| X95282.1 | CAA64603.1 | |||
| X97758.1 | CAA66352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834414 | Chr.2:150352860 - 150556795 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map