Assay Details
Target Gene Details
Entrez Gene ID: | 165055 |
Gene Name: | coiled-coil domain containing 138 |
Gene Aliases: |
- |
Location: |
Chr.2:108786730-108885485 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Gene Details
Entrez Gene ID: | 5903 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2 |
Location: |
Chr.2:108719446-109842301 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP2 | XM_017004623.1 | XP_016860112.1 |
More Information
Set Membership: |
|

Genomic Map