Assay Details
Target Gene Details
Entrez Gene ID: | 100529241 |
Gene Name: | HSPE1-MOB4 readthrough |
Gene Aliases: |
HSPE1-PHOCN |
Location: |
Chr.2:197499997-197553699 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPE1-MOB4 | NM_001202485.1 | NP_001189414.1 | ||
| BJ992310.1 | ||||
| CR990871.1 |
Target Gene Details
Entrez Gene ID: | 25843 |
Gene Name: | MOB family member 4, phocein |
Gene Aliases: |
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3 |
Location: |
Chr.2:197515571-197553699 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB4 | NM_001100819.2 | NP_001094289.1 | ||
| NM_001204094.1 | NP_001191023.1 | |||
| NM_015387.4 | NP_056202.2 | |||
| NM_199482.3 | NP_955776.1 | |||
| AB015441.1 | BAB19057.1 | |||
| AF093825.1 | AAP97221.1 | |||
| AF151853.1 | AAD34090.1 | |||
| AF250319.1 | AAG44567.1 | |||
| AJ580638.1 | CAE45270.1 | |||
| AK027043.1 | BAB15635.1 | |||
| AK289377.1 | ||||
| AK292938.1 | ||||
| AK297514.1 | ||||
| AK307497.1 | ||||
| AL080070.1 | CAB45697.1 | |||
| BC005237.1 | AAH05237.1 | |||
| CR457371.1 | CAG33652.1 | |||
| DC298610.1 | ||||
| DC350915.1 | ||||
| DC375649.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014881 | Chr.2:197432432 - 197668882 on Build GRCh38 | Gain |
|
| nsv979162 | Chr.2:197523680 - 197535381 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map